Assay Details
Gene Symbol: LOC5577645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838137.1 | XP_021693829.1 | 10 - 11 | 5259 | 84 | |
XM_021838138.1 | XP_021693830.1 | 10 - 11 | 5355 | 84 | ||
XM_021838139.1 | XP_021693831.1 | 10 - 11 | 5137 | 84 | ||
XM_021838140.1 | XP_021693832.1 | 10 - 11 | 5232 | 84 | ||
XM_021838141.1 | XP_021693833.1 | 10 - 11 | 5169 | 84 | ||
XM_021838142.1 | XP_021693834.1 | 10 - 11 | 5142 | 84 | ||
XM_021838143.1 | XP_021693835.1 | 10 - 11 | 4684 | 84 |
