Assay Details
Gene Symbol: LOC5577645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838139.1 | XP_021693831.1 | 1 - 2 | 73 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021838139.1 transcripts.
Set Membership: |
![]() ![]() |
