Assay Details
Gene Symbol: LOC5578803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854000.1 | XP_021709692.1 | 1 - 2 | 1426 | 95 | |
XM_021854002.1 | XP_021709694.1 | 2 - 3 | 585 | 95 | ||
XM_021854003.1 | XP_021709695.1 | 2 - 3 | 878 | 95 | ||
XM_021854004.1 | XP_021709696.1 | 2 - 3 | 454 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021854000.1, XM_021854002.1, XM_021854003.1, XM_021854004.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
