Assay Details
Gene Symbol: LOC5578803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854000.1 | XP_021709692.1 | 4 - 5 | 2954 | 63 | |
XM_021854002.1 | XP_021709694.1 | 5 - 6 | 2113 | 63 | ||
XM_021854003.1 | XP_021709695.1 | 5 - 6 | 2406 | 63 | ||
XM_021854004.1 | XP_021709696.1 | 5 - 6 | 1982 | 63 | ||
XM_021854005.1 | XP_021709697.1 | 4 - 5 | 1694 | 63 | ||
GenBank mRNA | L46373.1 | - | 3 - 4 | 1124 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
