Assay Details
Gene Symbol: LOC5578537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663878.2 | XP_001663928.2 | 6 - 7 | 2577 | 100 | |
XM_021841579.1 | XP_021697271.1 | 6 - 7 | 2600 | 100 | ||
XM_021841580.1 | XP_021697272.1 | 7 - 8 | 2534 | 100 | ||
XM_021841581.1 | XP_021697273.1 | 6 - 7 | 2038 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
