Assay Details
Gene Symbol: LOC5578537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841579.1 | XP_021697271.1 | 2 - 3 | 873 | 93 | |
XM_021841580.1 | XP_021697272.1 | 2 - 3 | 873 | 93 | ||
XM_021841581.1 | XP_021697273.1 | 2 - 3 | 311 | 93 |
