Assay Details
Gene Symbol: LOC110675293 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839796.1 | XP_021695488.1 | 1 - 2 | 155 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021839796.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
