Assay Details
Gene Symbol: LOC5575644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838049.1 | XP_021693741.1 | 4 - 5 | 3212 | 77 | |
XM_021838050.1 | XP_021693742.1 | 5 - 6 | 2891 | 77 | ||
XM_021838051.1 | XP_021693743.1 | 5 - 6 | 2336 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
