Assay Details
Gene Symbol: LOC5575644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838049.1 | XP_021693741.1 | 3 - 4 | 3079 | 60 | |
XM_021838050.1 | XP_021693742.1 | 4 - 5 | 2758 | 60 | ||
XM_021838051.1 | XP_021693743.1 | 4 - 5 | 2203 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
