Assay Details
Gene Symbol: LOC5578342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849011.1 | XP_021704703.1 | 2 - 3 | 520 | 73 | |
XM_021849012.1 | XP_021704704.1 | 2 - 3 | 476 | 73 | ||
XM_021849013.1 | XP_021704705.1 | 2 - 3 | 464 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021849011.1, XM_021849012.1, XM_021849013.1 transcripts.
Set Membership: |
![]() ![]() |
