Assay Details
Gene Symbol: LOC5578342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656861.2 | XP_001656911.2 | 3 - 4 | 2617 | 99 | |
XM_021849011.1 | XP_021704703.1 | 4 - 5 | 2865 | 99 | ||
XM_021849012.1 | XP_021704704.1 | 4 - 5 | 2821 | 99 | ||
XM_021849013.1 | XP_021704705.1 | 4 - 5 | 2809 | 99 |
