Assay Details
Gene Symbol: LOC5569185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652718.2 | XP_001652768.1 | 3 - 4 | 1152 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5569185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652718.2 | XP_001652768.1 | 3 - 4 | 1152 | 93 |
Set Membership: |
![]() ![]() ![]() |