Assay Details
Gene Symbol: LOC5569682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845097.1 | XP_021700789.1 | 6 - 7 | 3456 | 84 | |
XM_021845098.1 | XP_021700790.1 | 8 - 9 | 2546 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
