Assay Details
Gene Symbol: LOC5571510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848408.1 | XP_021704100.1 | 12 - 13 | 2914 | 92 | |
XM_021848409.1 | XP_021704101.1 | 12 - 13 | 2896 | 92 | ||
XM_021848410.1 | XP_021704102.1 | 11 - 12 | 2783 | 92 | ||
XM_021848411.1 | XP_021704103.1 | 12 - 13 | 3879 | 92 | ||
XM_021848412.1 | XP_021704104.1 | 12 - 13 | 2571 | 92 | ||
XM_021848414.1 | XP_021704106.1 | 12 - 13 | 2280 | 92 | ||
XM_021848415.1 | XP_021704107.1 | 12 - 13 | 2564 | 92 | ||
XM_021848416.1 | XP_021704108.1 | 12 - 13 | 2556 | 92 | ||
XM_021848417.1 | XP_021704109.1 | 12 - 13 | 2312 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
