Assay Details
Gene Symbol: LOC5571510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848408.1 | XP_021704100.1 | 4 - 5 | 1342 | 65 | |
XM_021848409.1 | XP_021704101.1 | 4 - 5 | 1324 | 65 | ||
XM_021848410.1 | XP_021704102.1 | 3 - 4 | 1211 | 65 | ||
XM_021848411.1 | XP_021704103.1 | 4 - 5 | 2307 | 65 | ||
XM_021848412.1 | XP_021704104.1 | 4 - 5 | 999 | 65 | ||
XM_021848414.1 | XP_021704106.1 | 4 - 5 | 708 | 65 | ||
XM_021848415.1 | XP_021704107.1 | 4 - 5 | 992 | 65 | ||
XM_021848416.1 | XP_021704108.1 | 4 - 5 | 984 | 65 | ||
XM_021848417.1 | XP_021704109.1 | 4 - 5 | 740 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
