Assay Details
Gene Symbol: LOC5571510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848408.1 | XP_021704100.1 | 7 - 8 | 1708 | 101 | |
XM_021848409.1 | XP_021704101.1 | 7 - 8 | 1690 | 101 | ||
XM_021848410.1 | XP_021704102.1 | 6 - 7 | 1577 | 101 | ||
XM_021848411.1 | XP_021704103.1 | 7 - 8 | 2673 | 101 | ||
XM_021848412.1 | XP_021704104.1 | 7 - 8 | 1365 | 101 | ||
XM_021848414.1 | XP_021704106.1 | 7 - 8 | 1074 | 101 | ||
XM_021848415.1 | XP_021704107.1 | 7 - 8 | 1358 | 101 | ||
XM_021848416.1 | XP_021704108.1 | 7 - 8 | 1350 | 101 | ||
XM_021848417.1 | XP_021704109.1 | 7 - 8 | 1106 | 101 |

More Information
Set Membership: |
![]() ![]() |
