Assay Details
Gene Symbol: LOC5571510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848408.1 | XP_021704100.1 | 8 - 9 | 2013 | 83 | |
XM_021848409.1 | XP_021704101.1 | 8 - 9 | 1995 | 83 | ||
XM_021848410.1 | XP_021704102.1 | 7 - 8 | 1882 | 83 | ||
XM_021848411.1 | XP_021704103.1 | 8 - 9 | 2978 | 83 | ||
XM_021848412.1 | XP_021704104.1 | 8 - 9 | 1670 | 83 | ||
XM_021848414.1 | XP_021704106.1 | 8 - 9 | 1379 | 83 | ||
XM_021848415.1 | XP_021704107.1 | 8 - 9 | 1663 | 83 | ||
XM_021848416.1 | XP_021704108.1 | 8 - 9 | 1655 | 83 | ||
XM_021848417.1 | XP_021704109.1 | 8 - 9 | 1411 | 83 |
