Assay Details
Gene Symbol: LOC5577066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840647.1 | XP_021696339.1 | 3 - 4 | 1022 | 64 | |
XM_021840649.1 | XP_021696341.1 | 4 - 5 | 668 | 64 | ||
XM_021840650.1 | XP_021696342.1 | 4 - 5 | 941 | 64 | ||
XM_021840651.1 | XP_021696343.1 | 4 - 5 | 755 | 64 | ||
XM_021840652.1 | XP_021696344.1 | 5 - 6 | 729 | 64 | ||
XM_021840653.1 | XP_021696345.1 | 4 - 5 | 800 | 64 | ||
XM_021840654.1 | XP_021696346.1 | 3 - 4 | 990 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
