Assay Details
Gene Symbol: LOC5571294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840149.1 | XP_021695841.1 | 5 - 6 | 4897 | 74 | |
XM_021840150.1 | XP_021695842.1 | 5 - 6 | 4897 | 74 | ||
XM_021840152.1 | XP_021695844.1 | 5 - 6 | 4897 | 74 | ||
XM_021840153.1 | XP_021695845.1 | 5 - 6 | 4897 | 74 | ||
XM_021840154.1 | XP_021695846.1 | 5 - 6 | 4897 | 74 |
