Assay Details
Gene Symbol: LOC5566057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853475.1 | XP_021709167.1 | 2 - 3 | 2666 | 79 | |
XM_021853476.1 | XP_021709168.1 | 2 - 3 | 2666 | 79 | ||
XM_021853477.1 | XP_021709169.1 | 2 - 3 | 2666 | 79 | ||
XM_021853478.1 | XP_021709170.1 | 2 - 3 | 2666 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
