Assay Details
Gene Symbol: LOC5566057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853475.1 | XP_021709167.1 | 1 - 2 | 2549 | 74 | |
XM_021853476.1 | XP_021709168.1 | 1 - 2 | 2549 | 74 | ||
XM_021853477.1 | XP_021709169.1 | 1 - 2 | 2549 | 74 | ||
XM_021853478.1 | XP_021709170.1 | 1 - 2 | 2549 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
