Assay Details
Gene Symbol: LOC5579654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648078.3 | XP_001648128.2 | 5 - 6 | 655 | 55 | |
XM_021848836.1 | XP_021704528.1 | 5 - 6 | 697 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
