Assay Details
Gene Symbol: LOC5579654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648078.3 | XP_001648128.2 | 4 - 5 | 529 | 87 | |
XM_021848836.1 | XP_021704528.1 | 4 - 5 | 571 | 87 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5579654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648078.3 | XP_001648128.2 | 4 - 5 | 529 | 87 | |
XM_021848836.1 | XP_021704528.1 | 4 - 5 | 571 | 87 |