Assay Details
Gene Symbol: LOC5568096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848303.1 | XP_021703995.1 | 8 - 9 | 2108 | 102 | |
XM_021848304.1 | XP_021703996.1 | 7 - 8 | 2036 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
