Assay Details
Gene Symbol: LOC5568096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848303.1 | XP_021703995.1 | 6 - 7 | 1201 | 64 | |
XM_021848304.1 | XP_021703996.1 | 5 - 6 | 1129 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
