Assay Details
Gene Symbol: LOC5568096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848303.1 | XP_021703995.1 | 1 - 2 | 99 | 72 | |
XM_021848304.1 | XP_021703996.1 | 1 - 2 | 99 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021848303.1, XM_021848304.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
