Assay Details
Gene Symbol: LOC5567930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651912.2 | XP_001651962.1 | 1 - 2 | 609 | 65 | |
XM_021843854.1 | XP_021699546.1 | 1 - 2 | 611 | 65 |

More Information
Set Membership: |
![]() ![]() |
