Assay Details
Gene Symbol: LOC5567930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651912.2 | XP_001651962.1 | 3 - 4 | 1196 | 62 | |
XM_021843854.1 | XP_021699546.1 | 3 - 4 | 1198 | 62 | ||
XM_021843867.1 | XP_021699559.1 | 3 - 4 | 1260 | 62 | ||
XM_021843868.1 | XP_021699560.1 | 3 - 4 | 707 | 62 |

More Information
Set Membership: |
![]() ![]() |
