Assay Details
Gene Symbol: LOC5567930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651912.2 | XP_001651962.1 | 7 - 8 | 1879 | 84 | |
XM_021843854.1 | XP_021699546.1 | 8 - 9 | 1920 | 84 | ||
XM_021843867.1 | XP_021699559.1 | 8 - 9 | 1982 | 84 | ||
XM_021843868.1 | XP_021699560.1 | 8 - 9 | 1429 | 84 |
