Assay Details
Gene Symbol: LOC5571904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659952.2 | XP_001660002.1 | 2 - 3 | 969 | 59 | |
XM_001659953.2 | XP_001660003.1 | 3 - 4 | 1189 | 59 | ||
XM_021855131.1 | XP_021710823.1 | 4 - 5 | 1648 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
