Assay Details
Gene Symbol: LOC5576958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842963.1 | XP_021698655.1 | 1 - 2 | 508 | 122 | |
XM_021842964.1 | XP_021698656.1 | 1 - 2 | 508 | 122 | ||
XM_021842965.1 | XP_021698657.1 | 1 - 2 | 508 | 122 | ||
XM_021842966.1 | XP_021698658.1 | 1 - 2 | 508 | 122 | ||
XM_021842967.1 | XP_021698659.1 | 1 - 2 | 508 | 122 | ||
XM_021842968.1 | XP_021698660.1 | 1 - 2 | 508 | 122 |

More Information
Set Membership: |
![]() ![]() |
