Assay Details
Gene Symbol: LOC5576764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662800.2 | XP_001662850.2 | 3 - 4 | 1891 | 117 | |
XM_021854253.1 | XP_021709945.1 | 3 - 4 | 1831 | 117 |

More Information
Set Membership: |
![]() ![]() |
