Assay Details
Gene Symbol: LOC5576764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662800.2 | XP_001662850.2 | 2 - 3 | 483 | 81 | |
XM_021854253.1 | XP_021709945.1 | 2 - 3 | 423 | 81 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5576764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662800.2 | XP_001662850.2 | 2 - 3 | 483 | 81 | |
XM_021854253.1 | XP_021709945.1 | 2 - 3 | 423 | 81 |