Assay Details
Gene Symbol: LOC5576411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656063.2 | XP_001656113.1 | 2 - 3 | 358 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001656063.2 transcripts.
Set Membership: |
![]() ![]() |
