Assay Details
Gene Symbol: LOC5578528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663869.2 | XP_001663919.2 | 4 - 5 | 1448 | 97 | |
XM_021855093.1 | XP_021710785.1 | 4 - 5 | 1481 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
