Assay Details
Gene Symbol: LOC5578528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663869.2 | XP_001663919.2 | 6 - 7 | 1832 | 78 | |
XM_021855093.1 | XP_021710785.1 | 6 - 7 | 1865 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
