Assay Details
Gene Symbol: LOC5567090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841756.1 | XP_021697448.1 | 8 - 9 | 3335 | 85 | |
XM_021841757.1 | XP_021697449.1 | 8 - 9 | 3267 | 85 | ||
XM_021841758.1 | XP_021697450.1 | 8 - 9 | 3320 | 85 | ||
XM_021841759.1 | XP_021697451.1 | 8 - 9 | 3252 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
