Assay Details
Gene Symbol: LOC5566024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650322.3 | XP_001650372.3 | 2 - 3 | 1383 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5566024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650322.3 | XP_001650372.3 | 2 - 3 | 1383 | 118 |
Set Membership: |
![]() ![]() ![]() |