Assay Details
Gene Symbol: LOC5578262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843982.1 | XP_021699674.1 | 1 - 2 | 1183 | 102 | |
XM_021843984.1 | XP_021699676.1 | 1 - 2 | 1183 | 102 | ||
XM_021843985.1 | XP_021699677.1 | 1 - 2 | 1183 | 102 | ||
XM_021843986.1 | XP_021699678.1 | 1 - 2 | 1183 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021843982.1, XM_021843984.1, XM_021843985.1, XM_021843986.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
