Assay Details
Gene Symbol: LOC5578262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843982.1 | XP_021699674.1 | 16 - 17 | 6806 | 69 | |
XM_021843983.1 | XP_021699675.1 | 16 - 17 | 5719 | 69 | ||
XM_021843984.1 | XP_021699676.1 | 16 - 17 | 6806 | 69 | ||
XM_021843985.1 | XP_021699677.1 | 16 - 17 | 6806 | 69 | ||
XM_021843986.1 | XP_021699678.1 | 16 - 17 | 6806 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
