Assay Details
Gene Symbol: LOC5569069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847340.1 | XP_021703032.1 | 2 - 3 | 884 | 70 | |
XM_021847341.1 | XP_021703033.1 | 3 - 4 | 958 | 70 | ||
XM_021847342.1 | XP_021703034.1 | 2 - 3 | 851 | 70 |

More Information
Set Membership: |
![]() ![]() |
