Assay Details
Gene Symbol: LOC5569069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847340.1 | XP_021703032.1 | 5 - 6 | 1257 | 80 | |
XM_021847341.1 | XP_021703033.1 | 6 - 7 | 1331 | 80 | ||
XM_021847342.1 | XP_021703034.1 | 5 - 6 | 1224 | 80 |
