Assay Details
Gene Symbol: LOC5569069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847340.1 | XP_021703032.1 | 7 - 8 | 1754 | 68 | |
XM_021847341.1 | XP_021703033.1 | 8 - 9 | 1828 | 68 | ||
XM_021847342.1 | XP_021703034.1 | 7 - 8 | 1721 | 68 |

More Information
Set Membership: |
![]() ![]() |
