Assay Details
Gene Symbol: LOC5564923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848061.1 | XP_021703753.1 | 10 - 11 | 1781 | 72 | |
XM_021848062.1 | XP_021703754.1 | 10 - 11 | 1811 | 72 | ||
XM_021848063.1 | XP_021703755.1 | 10 - 11 | 1758 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
