Assay Details
Gene Symbol: LOC5564923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848061.1 | XP_021703753.1 | 8 - 9 | 1535 | 89 | |
XM_021848062.1 | XP_021703754.1 | 8 - 9 | 1565 | 89 | ||
XM_021848063.1 | XP_021703755.1 | 8 - 9 | 1512 | 89 | ||
XM_021848065.1 | XP_021703757.1 | 8 - 9 | 1540 | 89 |
