Assay Details
Gene Symbol: LOC5564923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848061.1 | XP_021703753.1 | 3 - 4 | 582 | 66 | |
XM_021848062.1 | XP_021703754.1 | 3 - 4 | 612 | 66 | ||
XM_021848063.1 | XP_021703755.1 | 3 - 4 | 559 | 66 | ||
XM_021848065.1 | XP_021703757.1 | 3 - 4 | 587 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
