Assay Details
Gene Symbol: LOC5569699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848161.1 | XP_021703853.1 | 3 - 4 | 817 | 81 | |
XM_021848162.1 | XP_021703854.1 | 3 - 4 | 723 | 81 | ||
XM_021848163.1 | XP_021703855.1 | 3 - 4 | 612 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
