Assay Details
Gene Symbol: LOC5578735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851866.1 | XP_021707558.1 | 6 - 7 | 2004 | 99 | |
XM_021851867.1 | XP_021707559.1 | 7 - 8 | 2125 | 99 | ||
XM_021851868.1 | XP_021707560.1 | 6 - 7 | 1992 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
