Assay Details
Gene Symbol: LOC5578735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851866.1 | XP_021707558.1 | 4 - 5 | 1695 | 76 | |
XM_021851867.1 | XP_021707559.1 | 5 - 6 | 1816 | 76 | ||
XM_021851868.1 | XP_021707560.1 | 4 - 5 | 1683 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
