Assay Details
Gene Symbol: LOC5578735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851866.1 | XP_021707558.1 | 2 - 3 | 1373 | 73 | |
XM_021851867.1 | XP_021707559.1 | 3 - 4 | 1494 | 73 | ||
XM_021851868.1 | XP_021707560.1 | 2 - 3 | 1361 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
